Order Entry
Ireland
ContactUsLinkComponent
Human recombinant siglec-10 (from HEK293 cells), Biotin
Human recombinant siglec-10 (from HEK293 cells), Biotin
Supplier:  ACRO Biosystems
Certificates
Human recombinant siglec-10 (from HEK293 cells), Biotin
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Biotin
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His, Avitag™
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilised Biotinylated Human Siglec-10, His,Avitag (Cat. No. SI0-H82E3) at 5 μg/ml (100 μl/well) on streptavidin precoated (0,5 μg/well) plate, can bind Anti-Human Siglec-10 MAb, human IgG with a linear range of 0,6 - 20 ng/ml (QC tested).
  • Gene ID:
    Q96LC7-1
  • Protein synonyms:
    Siglec10|PRO940|SIGLEC10|SLG2|MGC126774
  • Protein/peptide name:
    Siglec-10
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    61,8 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS with Arginine, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together