Order Entry
Ireland
ContactUsLinkComponent
Human recombinant mucin-1 (from HEK293 cells)
Human recombinant mucin-1 (from HEK293 cells)
Supplier:  ACRO Biosystems
Certificates
Human recombinant mucin-1 (from HEK293 cells)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Gene ID:
    P15941-1
  • Protein synonyms:
    Mucin 1|Episialin|CA 15-3|PEMT|PUM|MUC-1/X|CD227|MUC-1/SEC|KL-6|EMA|MUC1|MAM6|PEM|H23AG|MUC1/ZD
  • Protein/peptide name:
    Mucin-1
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    21,3 kDa (partial α chain) and 8,4 kDa (partial β chain)
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together