Cynomolgus monkey recombinant serum albumin (from HEK293 cells)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Tag sequence:His Tag
- Storage conditions:−20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Biological activity:Cynomolgus serum albumin protein, His Tag (Cat. No. CSA-C52H4) immobilised on CM4 chip can bind cynomolgus/Rhesus macaque FCGRT&B2M heterodimer protein, His Tag&Strep II Tag (Cat. No. FCM-C5284) with an affinity constant of 0,235 μM as determined in a SPR assay (Biacore T200) (Routinely tested). Loaded biotinylated cynomolgus / Rhesus macaque FcRn Heterodimer protein, His,Avitag (SPR verified)(Cat. No. FCM-C82W3) on SA Biosensor, can bind cynomolgus serum albumin, His Tag(Cat. No. CSA-C52H4) with an affinity constant of 0,617 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:A2V9Z4-1
- Protein synonyms:Serum albumin|Alb|ALB
- Protein/peptide name:Serum Albumin
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:67,9 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Frequently Bought Together