Mouse recombinant SCF (from HEK293)
Catalog # ACRBSCF-M5228-20UG
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Pk:20 µG
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:−20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Gene ID:P20826
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:SCF|KL-1|Kitl|KL|SF|FPH2|KITLG|MGF|SHEP7
- Protein/peptide name:SCF
- Purity:>95%
- Molecular weight:19,5 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT