Mouse recombinant ROR2 (from HEK293)
Catalog # ACRBRO2-M5253100UG
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Pk:100 µG
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:−20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with glycine, arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Gene ID:Q9Z138-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:NTRKR2|ROR2
- Protein/peptide name:ROR2
- Purity:>90%
- Molecular weight:67,6 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with glycine, arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT