Mouse recombinant neuropilin-1 (from HEK293)
Catalog # ACRBNR1-M5254100UG
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Pk:100 µG
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:−20 °C
- Endotoxin content:Less than 0,1 EU per μg by the LAL method
- Biological activity:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised human VEGF165, Tag free (Cat. No. ACRBVE5-H4210) at 2 μg/ml (100 μl/well) can bind Mouse NRP1, Mouse IgG2a Fc Tag (Cat. No. ACRBNR1-M5254) with a linear range of 0,5 - 4 ng/ml (QC tested).
- Gene ID:P97333-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:NRP1|Neuropilin-1|VEGF165R|NRP|CD304
- Protein/peptide name:Neuropilin-1
- Purity:>90%
- Molecular weight:120,5 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT