Order Entry
Ireland
ContactUsLinkComponent
Human recombinant CD73 (from HEK293 cells)
Human recombinant CD73 (from HEK293 cells)
Supplier:  ACRO Biosystems
Certificates
Human recombinant CD73 (from HEK293 cells)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    Mouse IgG2a Fc Tag (active enzyme)
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 0,1 EU per μg by the LAL method.
  • Gene ID:
    P21589-1
  • Protein synonyms:
    5'-Nucleotidase|5'-NT|NTE|NT5|NT5E|CD73
  • Protein/peptide name:
    CD73
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    84,9 kDa
  • Endotoxin level:
    Low
  • Formulation:
    lyophilised from 0,22 μm filtered solution in 51 mM Tris, 100 mM Glycine, 150 mM NaCl, pH 7,5. Normally Trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together