Human recombinant LRRC32 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5, Normally trehalose is added as protectant before Lyophilisation,; Loaded Human LRRC32, Fc Tag (Cat, No, LR2-H5256) on AHC Biosensor, can bind Human TGFB1, His Tag (Cat, No, TG1-H524x) with an affinity constant of 0,288 μM as determined in BLI assay (ForteBio Octet Red96e) (QC tested).
- Gene ID:Q14392-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:GARP|D11S833E|Garpin
- Protein/peptide name:LRRC32
- Purity:>95%
- Molecular weight:92,4 kDa
- Sequence:01
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5, Normally trehalose is added as protectant before Lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together