4-Bromo-5-fluorobenzene-1,2-diamine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:153505-37-4
:
Formula:C₆H₆BrFN₂
MDL Number:MFCD08458150
Melting Pt:107 °C
:205.028 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together