Order Entry
Ireland
ContactUsLinkComponent
2,5-Difluoro-4-methylbenzoic acid methyl ester 95%
2,5-Difluoro-4-methylbenzoic acid methyl ester 95%
Catalog # APOSPC909715-1G
Supplier:  Apollo Scientific
CAS Number:  
2,5-Difluoro-4-methylbenzoic acid methyl ester 95%
Catalog # APOSPC909715-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :186.155 g/mol
Storage Temperature:Ambient
CAS Number:1355488-71-9
Formula:C₉H₈F₂O₂
MDL Number:MFCD16250137