
5-Chloro-6-fluoro-2,3-dihydro-1H-inden-1-one 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:881190-94-9
:
Formula:C₉H₆ClFO
MDL Number:MFCD13190260
:184.595 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together