Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Fluoro-5-hydroxy-4-methylbenzoic acid
2-Fluoro-5-hydroxy-4-methylbenzoic acid 95%
2-Fluoro-5-hydroxy-4-methylbenzoic acid 95%
APOSPC907093-5G
:
Apollo Scientific
:
2-Fluoro-5-hydroxy-4-methylbenzoic acid 95%
APOSPC907093-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
170.138 g/mol
:
Ambient
:
870221-14-0
:
C₈H₇FO₃
:
MFCD19440271