Order Entry
Ireland
ContactUsLinkComponent
5-Chloro-3-fluorobenzene-1,2-diol 95%
5-Chloro-3-fluorobenzene-1,2-diol 95%
Catalog # APOSPC907077-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Chloro-3-fluorobenzene-1,2-diol 95%
Catalog # APOSPC907077-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :162.546 g/mol
Storage Temperature:Ambient
CAS Number:645405-05-6
Formula:C₆H₄ClFO₂
MDL Number:MFCD18382509