
Ethyl 4-(4-fluorophenyl)-6-methyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5937-24-6
:
Formula:C₁₄H₁₅FN₂O₃
MDL Number:MFCD00500223
:278.279 g/mol
Melting Pt:181…185 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together