5-Bromo-2-fluorobenzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:339370-40-0
:
Formula:C₆H₃BrClFO₂S
MDL Number:MFCD09763690
:273.507 g/mol
Melting Pt:61…66 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
Frequently Bought Together