
4-Deoxy-4-fluoro-D-glucose
Catalog # APOSPC7308-25MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:182.147 g/mol
Melting Pt:189…190 °C
:GHS07
Storage Temperature:Freezer
CAS Number:29218-07-3
Formula:C₆H₁₁FO₅
MDL Number:MFCD00077529