1,2-Dihydro-1-(4-fluorophenyl)-2-oxopyridine-4-carbonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:929000-78-2
:
Formula:C₁₂H₇FN₂O
MDL Number:MFCD09258782
:214.195 g/mol
Storage Temperature:Ambient
Frequently Bought Together