
5-(Trifluoromethoxy)-1H-indazole, N1-BOC protected
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1346521-27-4
:
Formula:C₁₃H₁₃F₃N₂O₃
MDL Number:MFCD20481820
:302.249 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together