3-Bromo-2-fluorophenylacetylene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1701568-15-1
:
Formula:C₈H₄BrF
MDL Number:MFCD31654731
:199.02 g/mol
UN:1993
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together