
2-Bromo-5-fluorothiophenol 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:55389-14-5
:
Formula:C₆H₄BrFS
MDL Number:MFCD09475473
:207.063 g/mol
Boiling Pt:112…116 °C (35 mm Hg)
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together