(R)-6-Fluoro-2,3-dihydrobenzofuran-3-amine hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2102408-57-9
:
Formula:C₈H₈FNO·ClH
MDL Number:MFCD24435222
:189.615 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together