2-(Difluoromethyl)-5-methoxy-3-methyl-1H-indole
Catalog # APOSPC520071-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:211.208 g/mol
Storage Temperature:Ambient
CAS Number:2149590-55-4
Formula:C₁₁H₁₁F₂NO
MDL Number:MFCD29767555