2,4,6-Trichloro-5-(difluoromethyl)pyrimidine
Catalog # APOSPC520044-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:233.431 g/mol
Storage Temperature:Ambient
CAS Number:1807617-93-1
Formula:C₅HCl₃F₂N₂
MDL Number:MFCD29767527