Order Entry
Ireland
ContactUsLinkComponent
4-Oxo-1-[4-(trifluoromethyl)phenyl]cyclohexanecarboxylic acid
4-Oxo-1-[4-(trifluoromethyl)phenyl]cyclohexanecarboxylic acid
Catalog # APOSPC510127-5G
Supplier:  Apollo Scientific
CAS Number:  
4-Oxo-1-[4-(trifluoromethyl)phenyl]cyclohexanecarboxylic acid
Catalog # APOSPC510127-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :286.246 g/mol
Storage Temperature:Ambient
CAS Number:1385694-50-7
Formula:C₁₄H₁₃F₃O₃
MDL Number:MFCD22423132