3-Bromo-5-chloro-2-fluorobenzyl bromide 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2092620-40-9
:
Formula:C₇H₄Br₂ClF
MDL Number:MFCD31915765
:302.366 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
Frequently Bought Together