1,2-Diethynyl-4-fluorobenzene
Catalog # APOSPC50551-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:144.145 g/mol
Storage Temperature:Refrigerator
CAS Number:336106-79-7
Formula:C₁₀H₅F