Order Entry
Ireland
ContactUsLinkComponent
2,4,6-Trifluoroiodobenzene 95%
2,4,6-Trifluoroiodobenzene 95%
Catalog # APOSPC50367-1G
Supplier:  Apollo Scientific
CAS Number:  
2,4,6-Trifluoroiodobenzene 95%
Catalog # APOSPC50367-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :257.98 g/mol
Storage Temperature:Ambient
CAS Number:41860-63-3
Formula:C₆H₂F₃I
MDL Number:MFCD03095328