Order Entry
Ireland
ContactUsLinkComponent
 
5-Chloro-4-fluoro-2-methylaniline 97%
Catalog # APOSPC502149-250MG
Supplier:  Apollo Scientific
CAS Number:  
5-Chloro-4-fluoro-2-methylaniline 97%
Catalog # APOSPC502149-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :159.589 g/mol
Storage Temperature:Ambient
CAS Number:1242339-43-0
Formula:C₇H₇ClFN
MDL Number:MFCD16817674