4-Bromo-2-fluoro-3-methylbenzoic acid 95+%
Catalog # APOSPC501158-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:233.034 g/mol
Storage Temperature:Ambient
CAS Number:194804-90-5
Formula:C₈H₆BrFO₂
MDL Number:MFCD22052783