1,1'-Ethane-1,2-diylbis(2-fluorobenzene) 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:349-38-2
:
Formula:C₁₄H₁₂F₂
MDL Number:MFCD15143587
:218.242 g/mol
Melting Pt:40…41 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together