3-Bromo-4-fluorobenzene-1,2-diamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1257535-06-0
:
Formula:C₆H₆BrFN₂
MDL Number:MFCD18071059
:205.028 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together