Order Entry
Ireland
ContactUsLinkComponent
1H,1H,2H,3H,3H-Perfluoroheptane-1,2-diol 98%
1H,1H,2H,3H,3H-Perfluoroheptane-1,2-diol 98%
Catalog # APOSPC4785-1G
Supplier:  Apollo Scientific
CAS Number:  
1H,1H,2H,3H,3H-Perfluoroheptane-1,2-diol 98%
Catalog # APOSPC4785-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :294.115 g/mol
Boiling Pt:104…106 °C (5 mm Hg)
 :GHS07
Storage Temperature:Ambient
CAS Number:125070-38-4
Formula:C₇H₇F₉O₂
MDL Number:MFCD03424478