Order Entry
Ireland
ContactUsLinkComponent
1-Isobutyl-3-(trifluoromethyl)pyrazole
1-Isobutyl-3-(trifluoromethyl)pyrazole
Catalog # APOSPC448023-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Isobutyl-3-(trifluoromethyl)pyrazole
Catalog # APOSPC448023-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :192.182 g/mol
Storage Temperature:Ambient
CAS Number:1437795-15-7
Formula:C₈H₁₁F₃N₂
MDL Number:MFCD21668199