4-Bromo-1-isobutyl-3-(trifluoromethyl)pyrazole
Catalog # APOSPC448006-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:271.078 g/mol
Storage Temperature:Ambient
CAS Number:1437794-62-1
Formula:C₈H₁₀BrF₃N₂
MDL Number:MFCD24448782