3-Bromo-2-fluorobenzenesulphonyl chloride 95+%
Catalog # APOSPC446132-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:273.507 g/mol
Storage Temperature:Ambient
CAS Number:1214372-19-6
Formula:C₆H₃BrClFO₂S
MDL Number:MFCD13185362