[4-(Difluoromethyl)-6-(1,5-dimethyl-1H-pyrazol-4-yl)-3-methyl-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Catalog # APOSPC410486-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:335.309 g/mol
Storage Temperature:Ambient
CAS Number:1006477-98-0
Formula:C₁₅H₁₅F₂N₅O₂
MDL Number:MFCD08697730