Order Entry
Ireland
ContactUsLinkComponent
1-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-pyrrolidin-3-ylamine hydrochloride
1-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-pyrrolidin-3-ylamine hydrochloride
Catalog # APOSPC408412-500MG
Supplier:  Apollo Scientific
CAS Number:  
1-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-pyrrolidin-3-ylamine hydrochloride
Catalog # APOSPC408412-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :302.124 g/mol
Storage Temperature:Ambient
CAS Number:1420993-35-6
Formula:C₁₀H₁₁ClF₃N₃·ClH
MDL Number:MFCD24369845