(3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-3-ylmethyl)-carbamic acid tert-butyl ester
Catalog # APOSPC408395-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:393.832 g/mol
Storage Temperature:Ambient
CAS Number:1417793-53-3
Formula:C₁₇H₂₃ClF₃N₃O₂
MDL Number:MFCD22631676