Order Entry
Ireland
ContactUsLinkComponent
(S)-3-(4-Fluoro-benzenesulphonyl)-pyrrolidine-1-carboxylic acid tert-butyl ester
(S)-3-(4-Fluoro-benzenesulphonyl)-pyrrolidine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC408336-500MG
Supplier:  Apollo Scientific
CAS Number:  
(S)-3-(4-Fluoro-benzenesulphonyl)-pyrrolidine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC408336-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :329.387 g/mol
Storage Temperature:Ambient
CAS Number:1289585-25-6
Formula:C₁₅H₂₀FNO₄S
MDL Number:MFCD18837564