4-Hydroxy-2-n-propyl-6-trifluoromethylquinoline
Catalog # APOSPC407024-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:255.236 g/mol
Storage Temperature:Ambient
CAS Number:927800-87-1
Formula:C₁₃H₁₂F₃NO
MDL Number:MFCD02684130