Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Difluoromethoxy)pyridine-2-carbaldehyde
4-(Difluoromethoxy)pyridine-2-carbaldehyde 95%
4-(Difluoromethoxy)pyridine-2-carbaldehyde 95%
APOSPC400120-250MG
:
Apollo Scientific
:
4-(Difluoromethoxy)pyridine-2-carbaldehyde 95%
APOSPC400120-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
173.117 g/mol
:
Ambient
:
1315361-42-2
:
C₇H₅F₂NO₂
:
MFCD22552055