
6-Deoxy-6-fluoro-D-glucopyranose 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:34168-77-9
:
Formula:C₆H₁₁FO₅
MDL Number:MFCD00064096
:182.147 g/mol
Melting Pt:120…126 °C
Storage Temperature:Freezer
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together