Order Entry
Ireland
ContactUsLinkComponent
2-(2-Fluoroethoxy)-5-methylbenzoic acid 95%
2-(2-Fluoroethoxy)-5-methylbenzoic acid 95%
Catalog # APOSPC32902-100MG
Supplier:  Apollo Scientific
CAS Number:  
2-(2-Fluoroethoxy)-5-methylbenzoic acid 95%
Catalog # APOSPC32902-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :198.191 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1269797-08-1
Formula:C₁₀H₁₁FO₃
MDL Number:MFCD20661084