6-(Trifluoromethyl)-1,4,5,6-tetrahydropyrimidin-2-amine
Catalog # APOSPC32851-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:167.132 g/mol
Storage Temperature:Ambient
Formula:C₅H₈F₃N₃
MDL Number:MFCD28362447