Order Entry
Ireland
ContactUsLinkComponent
1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carboxaldehyde
Irritant
1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carboxaldehyde
Catalog # APOSPC31881-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carboxaldehyde
Catalog # APOSPC31881-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :204.2 g/mol
Melting Pt:111…117 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:423768-41-6
Formula:C₁₁H₉FN₂O
MDL Number:MFCD03407349