N1-(2,6-difluorophenyl)-2-chloroacetamide
Catalog # APOSPC31468-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:205.589 g/mol
Storage Temperature:Ambient
CAS Number:19064-26-7
Formula:C₈H₆ClF₂NO
MDL Number:MFCD00119514