3,5-Difluoro-4-methylbenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:103877-76-5
:
Formula:C₈H₆F₂O₂
MDL Number:MFCD20545741
Molecular Weight:172.129 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together