1-(4-Fluorophenyl)ethanone (2,4,6-trichlorophenyl)hydrazone
Catalog # APOSPC301016-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:331.6 g/mol
Storage Temperature:Ambient
CAS Number:394695-02-4
Formula:C₁₄H₁₀Cl₃FN₂
MDL Number:MFCD01132766