4-Bromo-1-(2,2-diethoxyethoxy)-2-fluorobenzene
Catalog # APOSPC300795-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:307.156 g/mol
Storage Temperature:Ambient
CAS Number:1271673-78-9
Formula:C₁₂H₁₆BrFO₃
MDL Number:MFCD19168379