Methyl 4-(2,6-dimethylmorpholin-4-yl)-2,5-difluorobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1858250-79-9
:
Formula:C₁₄H₁₇F₂NO₃
MDL Number:MFCD28103351
:285.286 g/mol
Storage Temperature:Ambient
Frequently Bought Together